missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human IFN-alpha 2 Protein

Product Code. 18667248 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18667248 100 μg 100µg
18646266 10 μg 10µg
18632127 500 μg 500µg
18633379 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18667248 Supplier Novus Biologicals™ Supplier No. NBP261318100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Fully biologically active when compared to standard. The activity is determined by the cytopathic effect inhibition assay.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS, pH 7.4, with 0.02 % Tween-20.
Gene ID (Entrez) 3440
Molecular Weight (g/mol) MolecularWeight-theroretical: 19.3 kDa
Quantity 100 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 0.1 EU/μg of IFN-alpha 2 AS determined by LAL method.
Gene Alias alpha-2 A interferon, IFNA, IFN-alpha-2, IFN-alphaA, INFA2, interferon alpha 2 b, interferon alpha A, interferon alpha-2, Interferon alpha-A, interferon, alpha 2, LeIF A, MGC125764, MGC125765
Gene Symbol IFNA2
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Research Category Apoptosis, Biologically Active Proteins, Cytokine Research
Purity or Quality Grade >97 % pure by SDS-PAGE and HPLC
Protein IFN-alpha 2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.