missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human IGF-I/IGF-1 LR3 Protein

Product Code. p-200083515 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
5 mg
20 mg
Unit Size:
1mg
20mg
5mg
This item is not returnable. View return policy

Product Code. 18640648

Brand: Novus Biologicals™ NBP2613145mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Assay 1: Fully biologically active when compared to standard. Measured in a serum-free cell proliferation assay using human MCF-7 cells. The ED50 for this effect is 0.3-1.5 ng/mL, corresponding to a specific activity of > 6.7 x 10^5 U/mg. Assay 2: Fully biologically active when compared to standard. The ED50 as determined by the stimulation of protein synthesis using rat L6 myoblasts is less than 10 ng/mL, corresponding to a specific activity of > 1.0 x 10^5 U/mg.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) SDS-PAGE, Bioactivity
Formulation Lyophilized from a 0.2 μm filtered concentrated solution in 20 mM PB, pH 7.2.
Gene ID (Entrez) 3479
Molecular Weight (g/mol) MolecularWeight-theroretical: 9.1 kDa
Quantity 5 mg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2 to 8 C for 1 month and (at -20°C to -70°C for long term storage. Avoid repeated freeze/thaw cycles.)
Endotoxin Concentration Less than 0.01 EU/μg of IGF-I/IGF-1 AS determined by LAL method.
Gene Alias IBP1, IGF1A, IGFI, IGF-I, IGF-IA, IGF-IB, insulin-like growth factor 1, insulin-like growth factor 1 (somatomedin C), insulin-like growth factor I, insulin-like growth factor IA, insulin-like growth factor IB, Mechano growth factor, MGF, somatomedin-C
Gene Symbol IGF1
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile distilled water or aqueous buffer containing 0.1% BSA to a concentration of 0.1-1.0 mg/mL Apportion stock solutions into working aliquots and store (at <-20°C.)
Research Category Biologically Active Proteins, Cell Cycle and Replication, Signal Transduction
Purity or Quality Grade >98%, by SDS-PAGE
Protein IGF-I/IGF-1
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado