missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human IGF-II/IGF2 Protein
Click to view available options
Quantity:
250 μg
50 μg
Unit Size:
250µg
50µg
Description
The Recombinant Human IGF-II/IGF2 Protein is derived from E. coli. The Recombinant Human IGF-II/IGF2 Protein has been validated for the following applications: Bioactivity.
Specifications
Specifications
| Formulation | Lyophilized from a 0.2μm filtered solution in Acetic Acid. |
| Gene ID (Entrez) | 3481 |
| Molecular Weight (g/mol) | 7.5 kDa |
| Name | IGF-II |
| Quantity | 50 μg |
| Source | E. coli-derived human IGF-II/IGF2 protein Ala25-Glu91 |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | C11orf43, chromosome 11 open reading frame 43, FLJ22066, FLJ44734, GRDF, IGF2, IGF-2, IGFII, IGF-II, insulin-like growth factor 2 (somatomedin A), insulin-like growth factor II, insulin-like growth factor type 2, MSA, PEG2, PP9974, somatomedin-A |
| Conjugate | Unconjugated |
| Recombinant | Recombinant |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction