missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IGF1 Protein

Product Code. 15914638
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 15914638

Brand: enQuireBio™ QP10680100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the IGF1 was constructed and used to recombinantly synthesize the protein.

Specifications

Gene ID (Entrez) 3479
Name IGF1 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol IGF1
Biological Activity The ED50 as determined by a cell proliferation assay using murine BALB/C 3T3 cells is less than 1.0 ng/ml, corresponding to a specific activity of > 1M 106 units/mg.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence GPETLCGAEL VDALQFVCGD RGFYFNKPTG YGSSSRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPAKSA
Buffer The protein was lyophilized from a 0.2microm filtered concentrated solution in 20mM PBS, pH 7.0, 130mM NaCl.
Purity or Quality Grade Greater than 98.0% as determined by:(a) Analysis by RP-HPLC(b) Analysis by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.