Learn More
enQuireBio™ Recombinant Human IGF1 Protein
Product Code. p-7148489
Click to view available options
Quantity:
1 mg
100 μg
500 μg
Unit Size:
100µg
1mg
500µg
Product Code. 15914638
Brand: enQuireBio™ QP10680100ug
A cDNA sequence encoding the IGF1 was constructed and used to recombinantly synthesize the protein.
Specifications
Gene ID (Entrez) 3479
Name IGF1 Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration < 1.0 EU per ug protein as determined by the LAL method.
Gene Symbol IGF1
Biological Activity The ED50 as determined by a cell proliferation assay using murine BALB/C 3T3 cells is less than 1.0 ng/ml, corresponding to a specific activity of > 1M 106 units/mg.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence GPETLCGAEL VDALQFVCGD RGFYFNKPTG YGSSSRRAPQ TGIVDECCFR SCDLRRLEMY CAPLKPAKSA
Buffer The protein was lyophilized from a 0.2microm filtered concentrated solution in 20mM PBS, pH 7.0, 130mM NaCl.
Purity or Quality Grade Greater than 98.0% as determined by:(a) Analysis by RP-HPLC(b) Analysis by SDS-PAGE.