missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human IL-2 Heat Stable Agonist Protein, CF

Código de producto. 30158289
missing translation for 'orderingAttributeHoverText'
Quantity:
1 mg
10 μg
100 μg
50 μg
500 μg
missing translation for 'unitSize'
100µg
10µg
1mg
500µg
50µg
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 30158289

missing translation for 'mfr': R&D Systems™ BT002HS050

Please to purchase this item. Need a web account? Register with us today!

Este artículo no se puede devolver. Vea la política de devoluciones

AI Modified Protein

Measured in a cell proliferation assay using NK-92 human natural killer lymphoma cells. The ED50 for this effect is 0.0150-0.150 ng/mL.
TRUSTED_SUSTAINABILITY

Tekniske data

Accession Number Engineered based on P60568.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 3558
Molecular Weight (g/mol) 7 kDa, under reducing conditions.
Quantity 50 μg
Source E. coli-derived human IL-2 protein Proprietary
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 3 months, -20 to -70 °C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method.
Gene Alias aldesleukin, IL2, IL-2, IL-2lymphokine, interleukin 2, interleukin-2, involved in regulation of T-cell clonal expansion, Proleukin, T cell growth factor, T-cell growth factor, TCGF
Conjugate Unconjugated
Reconstitution Reconstitute the 10 μg size at 100 μg/mL in PBS. Reconstitute all other sizes at 500 μg/mL in PBS.
Protein IL-2
Vis mere Vis mindre
Korrektion af produktindhold

Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.

Produkttitel

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.

Mange tak! Din feedback er blevet sendt. Hos Fisher Scientific arbejder vi altid på at forbedre vores indhold for dig. Vi sætter pris på din feedback.