missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL 28A Protein
Click to view available options
Quantity:
1 mg
10 μg
2 μg
Unit Size:
10µg
1mg
2µg
Specifications
Specifications
| Gene ID (Entrez) | 282616 |
| Name | IL 28A Protein |
| Quantity | 10 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | < 1.0 EU per ug protein as determined by the LAL method. |
| Gene Symbol | IFNL2 |
| Biological Activity | The specific activity was determined by the dose dependent protection of the cytopathic effect on A549 cells (human lung adenocarcinoma epithelial cell line) that were challenged with encephalomyocarditis (EMC) virus and is typically 0.5-5ng/ml. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | Human (HEK293) |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction