missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL-3 / Interleukin-3 Protein
A DNA sequence encoding the Homo sapiens (Human) IL-3 / Interleukin-3, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6220-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_000579.2 | |
IL-3 / Interleukin-3 Protein | |
Research Use Only | |
IL3 | |
Recombinant Protein | |
E. coli | |
APMTQTTPLK TSWVNCSNMI DEIITHLKQP PLPLLDFNNL NGEDQDILME NNLRRPNLEA FNRAVKSLQN ASAIESILKN LLPCLPLATA APTRHPIHIK DGDWNEFRRK LTFYLKTLEN AQAQQTTLSL AIF | |
Greater than 90% as determined by SDS-PAGE. |
3562 | |
50 μg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |