Learn More
DiscoverX Recombinant Human IL-6
Product Code. 17352599
Encompass
Change view
Click to view available options
Quantity:
20 μg
Unit Size:
20µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17352599 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 17352599 Supplier DiscoverX Supplier No. 921285
Recombinant Human IL-6 is an agonist for IL6R. Provided as 20μg of dry powder alongside reconstitution buffer.
Interleukin 6 (IL-6) is a pro-inflammatory cytokine protein and mediator of the immune response. Also acts as an anti-inflammatory myokine. Secreted by T cells, macrophages, osteoblasts, and smooth muscle cells for a variety of functions, including stimulating immune response for bacterial infection, burns, trauma, inducing fever, stimulating osteoclast formation, and inducing inflammation. Also inhibits TNF-alpha and IL-1 and activates IL-1ra and IL-10, leading to anti-inflammatory activity. Agonist of the IL-6 receptor, and implicated in multiple inflammatory diseases, including systemic juvenile rheumatoid arthritis and diabetes mellitus susceptibility. Provided as 20μg of dry powder alongside reconstitution buffer.
Specifications
Quantity 20 μg
Product Type Control Ligands