missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL-9 / Interleukin-9 Protein

Recombinant Protein

€ 4515.00 - € 7420.00

Specifications

Accession Number NP_000581.1
Gene ID (Entrez) 3578
Name IL-9 / Interleukin-9 Protein
Regulatory Status Research Use Only
Endotoxin Concentration Less than or equal to 1.00 EUs/ug as determined by the LAL method.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15936029
View Documents Promotion Details
enQuireBio™
QP5384-500UG
500 μg
€ 4515.00
500µg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15926029
View Documents Promotion Details
enQuireBio™
QP5384-1MG
1 mg
€ 7420.00
1mg
View alternative products
Save Up to 
»
Estimated Shipment: 28-05-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Specifications

NP_000581.1
IL-9 / Interleukin-9 Protein
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 2 ng/mL in the MO7e cell proliferation assay. The bioactivity acceptance criteria is set at 5 x 10^5 units/mg.
Human
Untagged
10 mM sodium phosphate, pH 7.5
3578
Research Use Only
IL9
Recombinant Protein
E. coli
MQGCPTLAGI LDINFLINKM QEDPASKCHC SANVTSCLCL GIPSDNCTRP CFSERLSQMT NTTMQTRYPL IFSRVKKSVE VLKNNKCPYF SCEQPCNQTT AGNALTFLKS LLEIFQKEKM RGMRGKI
Greater than 95% as determined by SDS-PAGE.
Videos
SDS
Documents

Documents

Product Certifications