missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL17D Protein
A cDNA sequence encoding the IL17D was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP10724-5ug
Additional Details : Weight : 0.01000kg
Specifications
53342 | |
5 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Immobilized rHuIL-17D binds to rHuIL-17BR with EC50 less than 2microg/ml. | |
Human | |
Untagged | |
Lyophilized from a 0.2microm filtered solution in Acetonitrile and TFA. |
IL17D Protein | |
Research Use Only | |
IL17D | |
Recombinant Protein | |
E. coli | |
APRAGRRPAR PRGCADRPEE LLEQLYGRLA AGVLSAFHHT LQLGPREQAR NASCPAGGRP ADRRFRPPTN LRSVSPWAYR ISYDPARYPR YLPEAYCLCR GCLTGLFGEE DVRFRSAPVY MPTVVLRRTP ACAGGRSVYT EAYVTIPVGC TCVPEPEKDA DSINSSIDKQ GAKLLLGPND APAGP | |
Greater than 97.0% as determined by SDS-PAGE. |