Learn More
enQuireBio™ Recombinant Human IL1F10 / IL-38 Protein
A DNA sequence encoding the Homo sapiens (Human) IL1F10 / IL-38, was expressed in the hosts and tags indicated.
Brand:  enQuireBio™ QP7890-ec-100ug
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 16011421
  • € 530.00 / 100µg
Estimated Shipment: 28-06-2024
to see stock.
Specifications
NP_115945.4
IL1F10 / IL-38 Protein
Research Use Only
Il1f10
Recombinant Protein
E. coli
MCSLPMARYY IIKYADQKAL YTRDGQLLVG DPVADNCCAE KICILPNRGL ARTKVPIFLG IQGGSRCLAC VETEEGPSLQ LEDVNIEELY KGGEEATRFT FFQSSSGSAF RLEAAAWPGW FLCGPAEPQQ PVQLTKESEP SARTKFYFEQ SW
Greater than 90% as determined by SDS-PAGE.
84639
100 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol