missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human IL2 / Interleukin-2 Protein

A DNA sequence encoding the Homo sapiens (Human) IL2 / Interleukin-2, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6216-ec-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031650

  • € 530.00 / 100µg
Estimated Shipment: 28-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_000577.2
IL2 / Interleukin-2 Protein
Research Use Only
IL2
Recombinant Protein
E. coli
APTSSSTKKT QLQLEHLLLD LQMILNGINN YKNPKLTRML TFKFYMPKKA TELKHLQCLE EELKPLEEVL NLAQSKNFHL RPRDLISNIN VIVLELKGSE TTFMCEYADE TATIVEFLNR WITFCQSIIS TLT
Greater than 90% as determined by SDS-PAGE.
3558
100 μg
Not determined.
Full Length
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers