missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human IL7R Protein
Click to view available options
Quantity:
1 μg
5 μg
50 μg
Unit Size:
1µg
50µg
5µg
Specifications
Specifications
| Name | IL7R Protein |
| Quantity | 5 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | Baculovirus |
| Protein Tag | His |
| Sequence | ADLESGYAQN GDLEDAELDD YSFSCYSQLE VNGSQHSLTC AFEDPDVNIT NLEFEICGAL VEVKCLNFRK LQEIYFIETK KFLLIGKSNI CVKVGEKSLT CKKIDLTTIV KPEAPFDLSV VYREGANDFV VTFNTSHLQK KYVKVLMHDV AYRQEKDENK WTHVNLSSTK LTLLQRKLQP AAMYEIKVRS IPDHYFKGFW SEWSPSYYFR TPEINNSSGE MDLEPKSCDK THTCPPCPAP ELLGGPSVFL FPPKPKDTLM ISRTPEVTCV VVDVSHEDPE VKFNWYVDGV EVHNAKTKPR EEQYNSTYRV VSVLTVLHQD WLNGKEYKCK VSNKALPAPI EKTISKAKGQ PREPQVYTLP PSRDELTKNQ VSLTCLVKGF YPSDIAVEWE SNGQPENNYK TTPPVLDSDG SFFLYSKLTV DKSRWQQGNV FSCSVMHEAL HNHYTQKSLS LSPGKHHHHH H |
| Buffer | IL7R protein solution (0.5 mg/ml) contains Phosphate Buffered Saline (pH 7.4) and 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction