missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ILKAP Protein

A DNA sequence encoding the Homo sapiens (Human) ILKAP, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6230-ec-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031780

Quantity Price
1 € 1180.00 / 500µg
3 € 1100.47 / 500µg
Estimated Shipment
02-11-2020
Add to basket


Specifications

Specifications

NP_110395.1
ILKAP Protein
500 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
80895
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ILKAP
Recombinant Protein
E. coli
MDLFGDLPEP ERSPRPAAGK EAQKGPLLFD DLPPASSTDS GSGGPLLFDD LPPASSGDSG SLATSISQMV KTEGKGAKRK TSEEEKNGSE ELVEKKVCKA SSVIFGLKGY VAERKGEREE MQDAHVILND ITEECRPPSS LITRVSYFAV FDGHGGIRAS KFAAQNLHQN LIRKFPKGDV ISVEKTVKRC LLDTFKHTDE EFLKQASSQK PAWKDGSTAT CVLAVDNILY IANLGDSRAI LCRYNEESQK HAALSLSKEH NPTQYEERMR IQKAGGNVRD GRVLGVLEVS RSIGDGQYKR CGVTSVPDIR RCQLTPNDRF ILLACDGLFK VFTPEEAVNF ILSCLEDEKI QTREGKSAAD ARYEAACNRL ANKAVQRGSA DNVTVMVVRI GH
Videos
SDS
Documents