missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Immunoglobulin lambda-like polypeptide 5 Protein

A DNA sequence encoding the Homo sapiens (Human) Immunoglobulin lambda-like polypeptide 5, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP9714-ye-500ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16028621

  • € 1740.00 / 500µg
Estimated Shipment: 12-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001171597.1
Immunoglobulin lambda-like polypeptide 5 Protein
Research Use Only
IGLL5
Recombinant Protein
Yeast
HGLLRPMVAP QSGDPDPGAS VGSSRSSLRS LWGRLLLQPS PQRADPRCWP RGFWSEPQSL CYVFGTGTKV TVLGQPKANP TVTLFPPSSE ELQANKATLV CLISDFYPGA VTVAWKADGS PVKAGVETTK PSKQSNNKYA ASSYLSLTPE QWKSHRSYSC QVTHEGSTVE KTVAPTECS
Greater than 90% as determined by SDS-PAGE.
100423062
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers