missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human INPP5A Protein
A DNA sequence encoding the Homo sapiens (Human) INPP5A, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7598-ec-500ug
Additional Details : Weight : 0.01000kg
Specifications
NP_001307971.1 | |
INPP5A Protein | |
Research Use Only | |
INPP5A | |
Recombinant Protein | |
E. coli | |
MAGKAAAPGT AVLLVTANVG SLFDDPENLQ KNWLREFYQV VHTHKPHFMA LHCQEFGGKN YEASMSHVDK FVKELLSSDA MKEYNRARVY LDENYKSQEH FTALGSFYFL HESLKNIYQF DFKAKKYRKV AGKEIYSDTL ESTPMLEKEK FPQDYFPECK WSRKGFIRTR WCIADCAFDL VNIHLFHDAS NLVAWETSPS VYSGIRHKAL GYVLDRIIDQ RFEKVSYFVF GDFNFRLDSK SVVETLCTKA TMQTVRAADT NEVVKLIFRE SDNDRKVMLQ LEKKLFDYFN QEVFRDNNGT ALLEFDKELS VFKDRLYELD ISFPPSYPYS EDARQGEQYM NTRCPAWCDR ILMSPSAKEL VLRSESEEKV VTYDHIGPNV CMGDHKPVFL AFRIMPGAGK PHAHVHKCC | |
Greater than 90% as determined by SDS-PAGE. |
3632 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |