missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human INPP5A Protein
A DNA sequence encoding the Homo sapiens (Human) INPP5A, was expressed in the hosts and tags indicated.
€ 160.00 - € 2385.00
Specifications
Accession Number | NP_001307971.1 |
---|---|
Gene ID (Entrez) | 3632 |
Name | INPP5A Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16089650
|
enQuireBio™
QP7598-EC-100UG |
100 μg |
€ 530.00
100µg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16009660
|
enQuireBio™
QP7598-EC-1MG |
1 mg |
€ 2385.00
1mg |
Estimated Shipment: 14-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16099650
|
enQuireBio™
QP7598-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16019660
|
enQuireBio™
QP7598-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16029660
|
enQuireBio™
QP7598-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16039660
|
enQuireBio™
QP7598-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_001307971.1 | |
INPP5A Protein | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3632 | |
Research Use Only | |
INPP5A | |
Recombinant Protein | |
E. coli | |
MAGKAAAPGT AVLLVTANVG SLFDDPENLQ KNWLREFYQV VHTHKPHFMA LHCQEFGGKN YEASMSHVDK FVKELLSSDA MKEYNRARVY LDENYKSQEH FTALGSFYFL HESLKNIYQF DFKAKKYRKV AGKEIYSDTL ESTPMLEKEK FPQDYFPECK WSRKGFIRTR WCIADCAFDL VNIHLFHDAS NLVAWETSPS VYSGIRHKAL GYVLDRIIDQ RFEKVSYFVF GDFNFRLDSK SVVETLCTKA TMQTVRAADT NEVVKLIFRE SDNDRKVMLQ LEKKLFDYFN QEVFRDNNGT ALLEFDKELS VFKDRLYELD ISFPPSYPYS EDARQGEQYM NTRCPAWCDR ILMSPSAKEL VLRSESEEKV VTYDHIGPNV CMGDHKPVFL AFRIMPGAGK PHAHVHKCC | |
Greater than 90% as determined by SDS-PAGE. |