missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Interferon alpha-21 Protein

A DNA sequence encoding the Homo sapiens (Human) Interferon alpha-21, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP6201-ye-100ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16031530

  • € 645.00 / 100µg
Estimated Shipment: 18-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_002166.2
Interferon alpha-21 Protein
Research Use Only
IFNA21
Recombinant Protein
Yeast
CDLPQTHSLG NRRALILLAQ MGRISPFSCL KDRHDFGFPQ EEFDGNQFQK AQAISVLHEM IQQTFNLFST KDSSATWEQS LLEKFSTELN QQLNDLEACV IQEVGVEETP LMNVDSILAV KKYFQRITLY LTEKKYSPCA WEVVRAEIMR SFSLSKIFQE RLRRKE
Greater than 90% as determined by SDS-PAGE.
3452
100 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers