missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human KCNJ10 Protein
A DNA sequence encoding the Homo sapiens (Human) KCNJ10, was expressed in the hosts and tags indicated.
€ 580.00 - € 1885.00
Specifications
Accession Number | NP_002232.2 |
---|---|
Gene ID (Entrez) | 3766 |
Name | KCNJ10 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16039091
|
enQuireBio™
QP9956-IV-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16049091
|
enQuireBio™
QP9956-IV-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16029091
|
enQuireBio™
QP9956-IV-100UG |
100 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_002232.2 | |
KCNJ10 Protein | |
Not determined. | |
Full Length Transmembrane Protein | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |
3766 | |
Research Use Only | |
KCNJ10 | |
Recombinant Protein | |
E. coli | |
MTSVAKVYYS QTTQTESRPL MGPGIRRRRV LTKDGRSNVR MEHIADKRFL YLKDLWTTFI DMQWRYKLLL FSATFAGTWF LFGVVWYLVA VAHGDLLELD PPANHTPCVV QVHTLTGAFL FSLESQTTIG YGFRYISEEC PLAIVLLIAQ LVLTTILEIF ITGTFLAKIA RPKKRAETIR FSQHAVVASH NGKPCLMIRV ANMRKSLLIG CQVTGKLLQT HQTKEGENIR LNQVNVTFQV DTASDSPFLI LPLTFYHVVD ETSPLKDLPL RSGEGDFELV LILSGTVEST SATCQVRTSY LPEEILWGYE FTPAISLSAS GKYIADFSLF DQVVKVASPS GLRDSTVRYG DPEKLKLEES LREQAEKEGS ALSVRISNV | |
Greater than 90% as determined by SDS-PAGE. |