missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human L-lactate dehydrogenase C chain Protein

Product Code. 16092220
Click to view available options
Quantity:
100 μg
20 μg
50 μg
Unit Size:
100µg
20µg
50µg
This item is not returnable. View return policy

Product Code. 16092220

Brand: enQuireBio™ QP6291ba100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) L-lactate dehydrogenase C chain, was expressed in the hosts and tags indicated.

Specifications

Accession Number NP_002292.1
Gene ID (Entrez) 3948
Name L-lactate dehydrogenase C chain Protein
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol LDHC
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species Baculovirus
Protein Tag His
Sequence STVKEQLIEK LIEDDENSQC KITIVGTGAV GMACAISILL KDLADELALV DVALDKLKGE MMDLQHGSLF FSTSKITSGK DYSVSANSRI VIVTAGARQQ EGETRLALVQ RNVAIMKSII PAIVHYSPDC KILVVSNPVD ILTYIVWKIS GLPVTRVIGS GCNLDSARFR YLIGEKLGVH PTSCHGWIIG EHGDSSVPLW SGVNVAGVAL KTLDPKLGTD SDKEHWKNIH KQVIQSAYEI IKLKGYTSWA IGLSVMDLVG SILKNLRRVH PVSTMVKGLY GIKEELFLSI PCVLGRNGVS DVVKINLNSE EEALFKKSAE TLWNIQKDLI F
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.