missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human LAS1L GST (N-Term) Protein

Product Code. 18457119 Shop All Bio Techne Products
missing translation for 'orderingAttributeHoverText'
Quantity:
2 μg
missing translation for 'unitSize'
2µg
This item is not returnable. View return policy

Product Code. 18457119

missing translation for 'mfr': Novus Biologicals™ H00081887P012ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 81887
Molecular Weight (g/mol) MolecularWeight-theroretical: 109.5 kDa
Quantity 2 μg
Storage Requirements Store at -80°C. Avoid freeze/thaw cycles.
Gene Alias dJ475B7.2, FLJ12525, FLJ30133, FLJ45274, LAS1-like (S. cerevisiae), protein LAS1 homolog
Gene Symbol LAS1L
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein LAS1L
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.