missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human LDLR Protein
Click to view available options
:
Recombinant Human LDL R Protein, Carrier-Free
Description
The Recombinant Human LDLR Protein is derived from HEK293. The Recombinant Human LDLR Protein has been validated for the following applications: Bioactivity.
Order Info
The product is shipped at ambient temperature.
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Name | LDLR |
| Source | Human embryonic kidney cell,HEK293-derived human LDL R protein Ala22-Arg788 (Asp193Ala),with a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
| Gene Alias | FH, FHC, LDL R, LDL receptor, LDLCQ2, low density lipoprotein receptor, low-density lipoprotein receptor, low-density lipoprotein receptor class A domain-containing protein 3 |
| Conjugate | Unconjugated |
For Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction