Learn More
enQuireBio™ Recombinant Human Leuprorelin Protein
Product Code. 15960799
Click to view available options
Quantity:
5 mg
25 mg
100 mg
Unit Size:
100mg
25mg
5mg
Product Code. 15960799
Brand: enQuireBio™ QP125555mg
Recombinant Protein
Specifications
Name Leuprorelin Protein
Quantity 5 mg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag Untagged
Sequence pGlu-His-Trp-Ser-Tyr-D-Leu-Leu-Arg-Pro-NHEt
Buffer The Leuprorelin was lyophilized from a concentrated (1 mg/ml) solution with no additives.
Purity or Quality Grade Greater than 98.0% as determined by RP-HPLC.