missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human LIN7C Protein

Product Code. 15910869
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
This item is not returnable. View return policy

Product Code. 15910869

Brand: enQuireBio™ QP125755ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A cDNA sequence encoding the LIN7C was constructed and used to recombinantly synthesize the protein.

Specifications

Name LIN7C Protein
Quantity 5 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His
Sequence MGCSHHHHHH SSGLVPRGSH MGSMAALGEP VRLERDICRA IELLEKLQRS GEVPPQKLQA LQRVLQSEFC NAVREVYEHV YETVDISSSP EVRANATAKA TVAAFAASEG HSHPRVVELP KTEEGLGFNI MGGKEQNSPI YISRIIPGGI ADRHGGLKRG DQLLSVNGVS VEGEHHEKAV ELLKAAQGKV KLVVRYTPKV LEEMESRFEK MRSAKRRQQT
Buffer LIN7C protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.2M NaCl and 30% glycerol.
Purity or Quality Grade Greater than 90.0% as determined by SDS-PAGE
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.