missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Lipolysis-stimulated lipoprotein receptor Protein

A DNA sequence encoding the Homo sapiens (Human) Lipolysis-stimulated lipoprotein receptor, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7800-ec-200ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16040881

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

Specifications

Specifications

NP_001247418.1
Lipolysis-stimulated lipoprotein receptor Protein
Research Use Only
LSR
Recombinant Protein
E. coli
MQQDGLGVGT RNGSGKGRSV HPSWPWCAPR PLRYFGRDAR ARRAQTAAMA LLAGGLSRGL GSHPAAAGRD AVVFVWLLLS TWCTAPARAI QVTVSNPYHV VILFQPVTLP CTYQMTSTPT QPIVIWKYKS FCRDRIADAF SPASVDNQLN AQLAAGNPGY NPYVECQDSV RTVRVVATKQ GNAVTLGDYY QGRRITITGN ADLTFDQTAW GDSGVYYCSV VSAQDLQGNN EAYAELIVLG RTSGVAELLP GFQAGPIED
Greater than 90% as determined by SDS-PAGE.
51599
200 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers