missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Livin alpha Protein

Product Code. 18116710
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18116710

Brand: R&D Systems™ 1161LV100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Inhibition Activity

Measured by its ability to reverse the inhibition of DEVD-AFC cleavage activity in cell extracts activated by addition of cytochrome c and dATP. The ED50 for this effect is 0.5-2.0 μM. Optimal dilutions should be determined by each laboratory for each application.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q96CA5
For Use With (Application) Inhibition Assay
Formulation Supplied as a 0.2 μm filtered solution in HEPES, NaCl, DTT and Glycerol.
Gene ID (Entrez) 79444
Molecular Weight (g/mol) M.W. (Observed): 38 kDa, reducing conditions; M.W. (theoretical): 34 kDa
Name Livin α/KIAP
Quantity 100 μg
Source E. coli-derived human Livin protein Met1-Ser298, with a C-terminal 6-His tag
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening.
Structural Form Monomer
Endotoxin Concentration <1.5 EU / 1 μg of the protein by the LAL method.
Gene Symbol BIRC7
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by silver stain.
Protein Livin
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.