missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Lutropin-choriogonadotropic hormone receptor Protein
A DNA sequence encoding the Homo sapiens (Human) Lutropin-choriogonadotropic hormone receptor, was expressed in the hosts and tags indicated.
€ 695.00 - € 1930.00
Specifications
Accession Number | NP_000224.2 |
---|---|
Gene ID (Entrez) | 3973 |
Name | Lutropin-choriogonadotropic hormone receptor Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952838
|
enQuireBio™
QP10085-MA-20UG |
20 μg |
€ 695.00
20µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942838
|
enQuireBio™
QP10085-MA-100UG |
100 μg |
€ 1930.00
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15962838
|
enQuireBio™
QP10085-MA-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_000224.2 | |
Lutropin-choriogonadotropic hormone receptor Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
3973 | |
Research Use Only | |
Lhcgr | |
Recombinant Protein | |
Mammal | |
EALCPEPCNC VPDGALRCPG PTAGLTRLSL AYLPVKVIPS QAFRGLNEVI KIEISQIDSL ERIEANAFDN LLNLSEILIQ NTKNLRYIEP GAFINLPRLK YLSICNTGIR KFPDVTKVFS SESNFILEIC DNLHITTIPG NAFQGMNNES VTLKLYGNGF EEVQSHAFNG TTLTSLELKE NVHLEKMHNG AFRGATGPKT LDISSTKLQA LPSYGLESIQ RLIATSSYSL KKLPSRETFV NLLEATLTYP SHCCAFRNLP TKEQNFSHSI SENFSKQCES TVRKVNNKTL YSSMLAESEL SGWDYEYGFC LPKTPRCAPE PDAFNPCEDI MGYDFL | |
Greater than 90% as determined by SDS-PAGE. |