missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Ly6D Fc Chimera Protein

Product Code. 18769136
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18769136

Brand: R&D Systems™ 10169L6050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ly6D (Lymphocyte antigen 6D), also known as E48 antigen, is a member of the lymphostromal cell membrane Ly6 protein superfamily

  • Recombinant Protein
  • Carrier Free (CF)
  • The carrier free version does not contain Bovine Serum Albumin (BSA)
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q14210
For Use With (Application) Bioactivity
Gene ID (Entrez) 8581
Molecular Weight (g/mol) 35 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived human Ly6D protein

Human Ly6D (Leu21-Asn98) Accession # Q14210GGGSHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 200 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.