missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Lysyl oxidase homolog 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Lysyl oxidase homolog 1, was expressed in the hosts and tags indicated.
€ 160.00 - € 2385.00
Specifications
Accession Number | NP_005567.2 |
---|---|
Gene ID (Entrez) | 4016 |
Name | Lysyl oxidase homolog 1 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16052410
|
enQuireBio™
QP6315-EC-100UG |
100 μg |
€ 530.00
100µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16072410
|
enQuireBio™
QP6315-EC-1MG |
1 mg |
€ 2385.00
1mg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16062410
|
enQuireBio™
QP6315-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16082410
|
enQuireBio™
QP6315-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16092410
|
enQuireBio™
QP6315-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16002420
|
enQuireBio™
QP6315-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_005567.2 | |
Lysyl oxidase homolog 1 Protein | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
4016 | |
Research Use Only | |
Loxl1 | |
Recombinant Protein | |
E. coli | |
VGSDTVRGQA RHPFGFGQVP DNWREVAVGD STGMARARTS VSQQRHGGSA SSVSASAFAS TYRQQPSYPQ QFPYPQAPFV SQYENYDPAS RTYDQGFVYY RPAGGGVGAG AAAVASAGVI YPYQPRARYE EYGGGEELPE YPPQGFYPAP ERPYVPPPPP PPDGLDRRYS HSLYSEGTPG FEQAYPDPGP EAAQAHGGDP RLGWYPPYAN PPPEAYGPPR ALEPPYLPVR SSDTPPPGGE RNGAQQGRLS VGSVYRPNQN GRGLPDLVPD PNYVQASTYV QRAHLYSLRC AAEEKCLAST AYAPEATDYD VRVLLRFPQR VKNQGTADFL PNRPRHTWEW HSCHQHYHSM DEFSHYDLLD AATGKKVAEG HKASFCLEDS TCDFGNLKRY ACTSHTQGLS PGCYDTYNAD IDCQWIDITD VQPGNYILKV HVNPKYIVLE SDFTNNVVRC NIHYTGRYVS ATNCKIVQS | |
Greater than 90% as determined by SDS-PAGE. |