missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Major surface glycoprotein 1

Product Code. 16068740
Change view
Click to view available options
Quantity:
1 mg
10 μg
100 μg
200 μg
50 μg
500 μg
Unit Size:
100µg
10µg
1mg
200µg
500µg
50µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16068740 100 μg 100µg
16078740 10 μg 10µg
16018750 50 μg 50µg
16098740 200 μg 200µg
16008750 500 μg 500µg
16088740 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16068740 Supplier enQuireBio™ Supplier No. QP7412ec100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A DNA sequence encoding the Homo sapiens (Human) Major surface glycoprotein G, was expressed in the hosts and tags indicated.

Specifications

Name Major surface glycoprotein 1
Quantity 100 μg
Regulatory Status Research Use Only
Endotoxin Concentration Not determined.
Gene Symbol G
Protein Length Full Length
Product Type Recombinant Protein
Cross Reactivity Human
Species E. coli
Protein Tag His-SUMO
Sequence HKVTPTTAII QDATSQIKNT TPTYLTQNPQ LGISPSNPSE ITSQITTILA STTPGVKSTL QSTTVKTKNT TTTQTQPSKP TTKQRQNKPP SKPNNDFHFE VFNFVPCSIC SNNPTCWAIC KRIPNKKPGK KTTTKPTKKP TLKTTKKDPK PQTTKSKEVP TTKPTEEPTI NTTKTNIITT LLTSNTTGNP ELTSQMETFH STSSEGNPSP SQVSTTSEYP SQPSSPPNTP RQ
Buffer Tris-based buffer, 50% glycerol
Purity or Quality Grade Greater than 90% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.