missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MAP1LC3A Protein
A cDNA sequence encoding the MAP1LC3A was constructed and used to recombinantly synthesize the protein.
Brand: enQuireBio™ QP12634-20ug
Additional Details : Weight : 0.01000kg
Specifications
MAP1LC3A Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MPSDRPFKQR RSFADRCKEV QQIRDQHPSK IPVIIERYKG EKQLPVLDKT KFLVPDHVNM SELVKIIRRR LQLNPTQAFF LLVNQHSMVS VSTPIADIYE QEKDEDGFLY MVYASQETFG LEHHHHHH | |
Greater than 95.0% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The MAP1LC3A solution (1 mg/ml) contains 20mM Tris-HCl pH 8.0, 1mM DTT, 0.1M NaCl and 20% glycerol. |