missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human MAT2B Protein

Product Code. 18216613 Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.1mg
This item is not returnable. View return policy

Product Code. 18216613

Brand: Novus Biologicals™ NBP130288

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: SDS-Page

An un-tagged recombinant protein corresponding to the amino acids 1-323 of Human MAT2B The Recombinant Human MAT2B Protein is derived from E. coli. The Recombinant Human MAT2B Protein has been validated for the following applications: SDS-Page.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9NZL9
For Use With (Application) ELISA, Immunoprecipitation, SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0), 1mM DTT, 10% glycerol
Gene ID (Entrez) 27430
Molecular Weight (g/mol) 36.4kDa
Name MAT2B Protein
Purification Method Protein
Quantity 0.1 mg
Source E. Coli
Immunogen MAT II beta, 1-323aa, Human, E.coli (AAH05218).
Storage Requirements −80°C. Avoid freeze-thaw cycles.
Regulatory Status RUO
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Purity or Quality Grade >90%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.