missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MED1 Protein
A DNA sequence encoding the Homo sapiens (Human) MED1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7584-ec-50ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004765.2 | |
MED1 Protein | |
Research Use Only | |
MED1 | |
Recombinant Protein | |
E. coli | |
FGEEYFDESS QSGDNDDFKG FASQALNTLG VPMLGGDNGE TKFKGNNQAD TVDFSIISVA GKALAPADLM EHHSGSQGPL LTTGDLGKEK TQKRVKEGNG TSNSTLSGPG LDSKPGKRSR TPSNDGKSKD KPPKRKKADT EGKSPSHSSS NRPF | |
Greater than 90% as determined by SDS-PAGE. |
5469 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |