missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Mediator of DNA damage checkpoint protein 1

A DNA sequence encoding the Homo sapiens (Human) Mediator of DNA damage checkpoint protein 1, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7613-ye-50ug

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16029790

Quantity Price
1 € 416.00 / 50µg
6 € 388.50 / 50µg
Estimated Shipment
03-11-2020
Add to basket


Specifications

Specifications

NP_055456.2
Mediator of DNA damage checkpoint protein 1
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
9656
Greater than 90% as determined by SDS-PAGE.
Research Use Only
MDC1
Recombinant Protein
Yeast
APKVLFTGVV DARGERAVLA LGGSLAGSAA EASHLVTDRI RRTVKFLCAL GRGIPILSLD WLHQSRKAGF FLPPDEYVVT DPEQEKNFGF SLQDALSRAR ERRLLEGYEI YVTPGVQPPP PQMGEIISCC GGTYLPSMPR SYKPQRVVIT CPQDFPHCSI PLRVGLPLLS PEFLLTGVLK QEAKPEAFVL S
Videos
SDS
Documents