missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human MICA His Protein

Product Code. 18416521 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
10 μg
100 μg
500 μg
Unit Size:
100µg
10µg
1mg
500µg
This item is not returnable. View return policy

Product Code. 18416521

Brand: Novus Biologicals™ NBP234993100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

MICA Protein is fully biologically active when compared to standard. The specific activity is determined by binding MICA antibody in ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Concentration LYOPH
For Use With (Application) PAGE, Bioactivity
Formulation 0.2 um filtered concentrated sterile solution in PBS, pH 7.4, and 8 M Urea.
Gene ID (Entrez) 100507436
Molecular Weight (g/mol) 36.9 kDa
Quantity 100 μg
Storage Requirements Store at -20°C to -70°C as supplied. After reconstitution, store at 2°C to 8°C for 1 month and at -20°C to -70°C for long term storage. Avoid repeated freeze-thaw cycles.
Endotoxin Concentration Less than 1 EU/ug of MICA as determined by LAL method.
Gene Alias FLJ60820, MGC111087, PERB11.1
Gene Symbol MICA
Reconstitution Recommended to centrifuge prior to opening. Reconstitute in sterile dH2O to a conc. of 1.0mg/mL. Apportion stock solutions into working aliquots and store at <-20°C. Make further dilutions in appropriate buffered solutions, with 8M Urea.
Research Category Biologically Active Proteins
Purity or Quality Grade >95% by SDS-PAGE and HPLC
Protein MICA
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.