missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Recombinant Human MIER3 GST (N-Term) Protein

Product Code. 18518280 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
10 μg
25 μg
Unit Size:
10µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18518280 10 μg 10µg
18547300 25 μg 25µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18518280 Supplier Novus Biologicals™ Supplier No. H00166968P0110ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

This protein was produced in an in vitro wheat germ expression system that should preserve correct conformational folding that is necessary for biological function. While it is possible that this protein could display some level of activity, the functionality of this protein has not been explicitly measured or validated.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Western Blot, ELISA, Protein Array, Immunoaffinity Purification
Formulation 50 mM Tris-HCl, 10 mM reduced Glutathione, pH 8.0 in the elution buffer.
Gene ID (Entrez) 166968
Molecular Weight (g/mol) MolecularWeight-theroretical: 84.9 kDa
Quantity 10 μg
Storage Requirements Store at -80°C. Avoid freeze/thaw cycles.
Gene Alias DKFZp686L09111, DKFZp781G1119, DKFZp781I1119, FLJ35954, mesoderm induction early response 1, family member 3, mesoderm induction early response protein 3, mi-er3
Gene Symbol MIER3
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Protein MIER3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.