missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human MKRN3 Protein
A DNA sequence encoding the Homo sapiens (Human) MKRN3, was expressed in the hosts and tags indicated.
€ 202.00 - € 2840.00
Specifications
Accession Number | NP_005655.1 |
---|---|
Gene ID (Entrez) | 7681 |
Name | MKRN3 Protein |
Regulatory Status | Research Use Only |
Endotoxin Concentration | Not determined. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16009370
|
enQuireBio™
QP7557-EC-100UG |
100 μg |
€ 655.00
100µg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16029370
|
enQuireBio™
QP7557-EC-1MG |
1 mg |
€ 2840.00
1mg |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16019370
|
enQuireBio™
QP7557-EC-10UG |
10 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16039370
|
enQuireBio™
QP7557-EC-200UG |
200 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16049370
|
enQuireBio™
QP7557-EC-500UG |
500 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
16059370
|
enQuireBio™
QP7557-EC-50UG |
50 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
NP_005655.1 | |
MKRN3 Protein | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |
7681 | |
Research Use Only | |
MKRN3 | |
Recombinant Protein | |
E. coli | |
MEEPAAPSEA HEAAGAQAGA EAAREGVSGP DLPVCEPSGE SAAPDSALPH AARGWAPFPV APVPAHLRRG GLRPAPASGG GAWPSPLPSR SSGIWTKQII CRYYIHGQCK EGENCRYSHD LSGRKMATEG GVSPPGASAG GGPSTAAHIE PPTQEVAEAP PAASSLSLPV IGSAAERGFF EAERDNADRG AAGGAGVESW ADAIEFVPGQ PYRGRWVASA PEAPLQSSET ERKQMAVGSG LRFCYYASRG VCFRGESCMY LHGDICDMCG LQTLHPMDAA QREEHMRACI EAHEKDMELS FAVQRGMDKV CGICMEVVYE KANPNDRRFG ILSNCNHSFC IRCIRRWRSA RQFENRIVKS CPQCRVTSEL VIPSEFWVEE EEEKQKLIQQ YKEAMSNKAC RYFAEGRGNC PFGDTCFYKH EYPEGWGDEP PGPGGGSFSA YWHQLVEPVR MGEGNMLYKS IKKELVVLRL ASLLFKRFLS LRDELPFSED QWDLLHYELE EYFNLIL | |
Greater than 90% as determined by SDS-PAGE. |