missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human MMP-14/MT1-MMP (NS0-expressed) Protein
Click to view available options
Quantity:
10 μg
Unit Size:
10µg
Description
Measured by its ability to cleave a fluorogenic peptide substrate Mca-KPLGL-Dpa-AR-NH2 (Catalog # ES010 ). The specific activity is >350 pmol/min/μg, as measured under the described conditions.
Specifications
Specifications
| Accession Number | P50281 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Sodium Acetate, NaCl, CaCl2, Glycerol. |
| Gene ID (Entrez) | 4323 |
| Molecular Weight (g/mol) | M.W. (Observed): 60-65 kDa, reducing conditions; M.W. (theoretical): 60 kDa |
| Name | MMP-14/MT1-MMP |
| Quantity | 10 μg |
| Source | Mouse myeloma cell line, NS0-derived human MMP-14/MT1-MMP protein Ala21-Ser538, with an Arg108Pro mutation and a C-terminal 6-His tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction