missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human/Mouse FGF-8b Protein

Product Code. 18726268
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 18726268

Brand: R&D Systems™ 423F801M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity

The Recombinant Human/Mouse FGF-8b Protein is derived from E. coli. The Recombinant Human/Mouse FGF-8b Protein has been validated for the following applications: Bioactivity.

Order Info

Ships at ambient temperature.

TRUSTED_SUSTAINABILITY

Specifications

Accession Number NP_006110 (human), NP_001159834 (mouse)
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in MOPS, Na 2 SO 4 and Brij-35 with BSA as a carrier protein.
Gene ID (Entrez) 2253
Molecular Weight (g/mol) 22.5 kDa
Quantity 1 mg
Source E. coli-derived FGF-8 protein Gln23-Arg215,with an N-terminal Met
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 3 months, 2 to 8° C under sterile conditions after reconstitution.
Regulatory Status RUO
Endotoxin Concentration <0.01 EU per 1μg of the protein by the LAL method
Gene Alias AIGF, AIGFKAL6
Gene Symbol FGF8
Biological Activity 6.5 to 40ng/mL
Product Type Recombinant protein
Conjugate Unconjugated
Species Human, Mouse
Recombinant Recombinant
Purity or Quality Grade >97%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Show More Show Less

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.