missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human/Mouse/Rat UbcH5c/UBE2D3 Protein

Product Code. 18746858
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
This item is not returnable. View return policy

Product Code. 18746858

Brand: R&D Systems™ E2627100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity

Recombinant Human UbcH5c/UBE2D3 is a member of the Ubiquitin-conjugating (E2) enzyme family that receives Ubiquitin from a Ubiquitin-activating (E1) enzyme and subsequently interacts with a Ubiquitin ligase (E3) to conjugate Ubiquitin to substrate proteins. Reaction conditions will need to be optimized for each specific application. We recommend an initial Recombinant Human UbcH5c/UBE2D3 concentration of 0.1-1 μM.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P61077.1
For Use With (Application) Enzyme Activity
Formulation Supplied as a 0.2 μm filtered solution in HEPES, NaCl, TCEP and Glycerol.
Gene ID (Entrez) 7323
Molecular Weight (g/mol) M.W. (theoretical): 17 kDa
Name UbcH5c/UBE2D3
Quantity 100 μg
Source E. coli-derived UbcH5c/UBE2D3 protein Met1 - Met147
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening.
Gene Symbol UBE2D3
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain.
Protein UbcH5c/UBE2D3
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.