missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human MSRB3 Protein

A DNA sequence encoding the Homo sapiens (Human) MSRB3, was expressed in the hosts and tags indicated.

Brand:  enQuireBio™ QP7810-ec-1mg

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 16030941

  • € 2385.00 / 1mg
Estimated Shipment: 17-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

NP_001026849.1
MSRB3 Protein
Research Use Only
MSRB3
Recombinant Protein
E. coli
MSAFNLLHLV TKSQPVALRA CGLPSGSCRD KKNCKVVFSQ QELRKRLTPL QYHVTQEKGT ESAFEGEYTH HKDPGIYKCV VCGTPLFKSE TKFDSGSGWP SFHDVINSEA ITFTDDFSYG MHRVETSCSQ CGAHLGHIFD DGPRPTGKRY CINSAALSFT PADSSGTAEG GSGVASPAQA DKAEL
Greater than 80% as determined by SDS-PAGE.
253827
1 mg
Not determined.
Full Length of Isoform 2
Human
GST
Tris-based buffer, 50% glycerol
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers