missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Myosin-binding protein C, fast-type Protein
A DNA sequence encoding the Homo sapiens (Human) Myosin-binding protein C, fast-type, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP7562-ec-10ug
Additional Details : Weight : 0.01000kg
Specifications
NP_004524.3 | |
Myosin-binding protein C, fast-type Protein | |
Research Use Only | |
MYBPC2 | |
Recombinant Protein | |
E. coli | |
EPLHLIVEDV TDTTTTLKWR PPNRIGAGGI DGYLVEYCLE GSEEWVPANT EPVERCGFTV KNLPTGARIL FRVVGVNIAG RSEPATLAQP VTIREIAEPP KIRLPRHLRQ TYIRKVGEQL NLVVPFQGKP RPQVVWTKGG APLDTSRVHV RTSDFDTVFF VRQAARSDSG EYELSVQIEN MKDTATIRIR VVEKAGPPIN VMVKEVWGTN ALVEWQAPKD DGNSEIMGYF VQKADKKTME WFNVYERNRH TSCTVSDLIV GNEYYFRVYT ENICGLSDSP GVSKNTARIL KTGITFKPFE YKEHDFRMAP KFLTPLIDRV VVAGYSAALN CAVRGHPKPK VVWMKNKMEI REDPKFLITN YQGVLTLNIR RPSPFDAGTY TCRAVNELGE ALAECKLEVR VPQ | |
Greater than 90% as determined by SDS-PAGE. |
4606 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |