missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human N-Acetylglucosaminyltransferase2/MGAT2
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
Description
Measured by its ability to modify the glycan Cy5-labeled M1N1f and thereby creating a band shift. Able to convert >50% of the substrate glycan M1N1f to the product glycan, as measured under the described conditions.
Specifications
Specifications
| Accession Number | Q10469.1 |
| For Use With (Application) | Enzyme Activity |
| Formulation | Supplied as a 0.2 μm filtered solution in Tris and NaCl. |
| Gene ID (Entrez) | 4247 |
| Molecular Weight (g/mol) | M.W.-theroretical: 48.7 kDa, M.W.-observed: 46-54 kDa, under reducing conditions |
| Quantity | 25 μg |
| Source | Human embryonic kidney cell, HEK293-derived human N-Acetylglucosaminyltransferase 2/MGAT2 protein Asn34-Gln447, with N-terminal HA tag |
| Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Endotoxin Concentration | <1.0 EU / 1 μg of the protein by the LAL method. |
| Gene Alias | alpha-1,6-mannosyl-glycoprotein 2-beta-N-acetylglucosaminyltransferase, Beta-1,2-N-acetylglucosaminyltransferase II, CDG2A, EC 2.4.1.143, GlcNAc-T II, GLCNACTII, GNT2, GNT-IICDGS2, Mannoside acetylglucosaminyltransferase 2, mannosyl (alpha-1,6-)-glycoprotein beta-1,2-N-acetylglucosaminyltransferase, N-glycosyl-oligosaccharide-glycoprotein N-acetylglucosaminyltransferase II, UDP-N-acetylglucosamine:alpha-6-D-mannosidebeta-1,2-N-acetylglucosaminyltransferase II |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction