missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NAGA Protein
Click to view available options
Quantity:
1 μg
5 μg
50 μg
Unit Size:
1µg
50µg
5µg
Specifications
Specifications
| Name | NAGA Protein |
| Quantity | 5 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | Baculovirus |
| Protein Tag | His |
| Sequence | LDNGLLQTPP MGWLAWERFR CNINCDEDPK NCISEQLFME MADRMAQDGW RDMGYTYLNI DDCWIGGRDA SGRLMPDPKR FPHGIPFLAD YVHSLGLKLG IYADMGNFTC MGYPGTTLDK VVQDAQTFAE WKVDMLKLDG CFSTPEERAQ GYPKMAAALN ATGRPIAFSC SWPAYEGGLP PRVNYSLLAD ICNLWRNYDD IQDSWWSVLS ILNWFVEHQD ILQPVAGPGH WNDPDMLLIG NFGLSLEQSR AQMALWTVLA APLLMSTDLR TISAQNMDIL QNPLMIKINQ DPLGIQGRRI HKEKSLIEVY MRPLSNKASA LVFFSCRTDM PYRYHSSLGQ LNFTGSVIYE AQDVYSGDII SGLRDETNFT VIINPSGVVM WYLYPIKNLE MSQQHHHHHH |
| Buffer | NAGA protein solution (0.5 mg/ml) containing Phosphate buffered saline (pH 7.4), 10% glycerol. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction