missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human NCAM-1/CD56 Fc Chimera Protein

Product Code. 18767136
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
This item is not returnable. View return policy

Product Code. 18767136

Brand: R&D Systems™ 10136NC050

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Neural cell adhesion molecule 1 (NCAM-1), also known as CD56, is a multifunctional member of the Ig superfamily

  • It belongs to a family of membrane-bound glycoproteins that are involved in Ca++ independent cell matrix and homophilic or heterophilic cell-cell interactions.
  • NCAM-1 in the adult brain shows a decline of sialylation relative to earlier developmental periods.
  • In regions that retain a high degree of neuronal plasticity, however, the adult brain continues to express polysialylated-NCAM-1, suggesting sialylation of NCAM-1 is involved in regenerative processes and synaptic plasticity
  • NCAM can function as a receptor for GDNF family of ligands. GDNF stimulates Schwann cell migration and neurite outgrowth in hippocampal and cortical neurons through binding to NCAM-1 and activation of Fyn, independently of RET
  • NCAM-1 also interacts with Ig IIIC isoform of the FGFR1 and plays an important role in developmental events as well as tumor progression
  • Due to expression on immune cells NCAM-1 has great implications on tumor progression, infection, and disease development
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P13591-1
For Use With (Application) Bioactivity
Gene ID (Entrez) 4684
Molecular Weight (g/mol) 103 kDa
Purification Method >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining
Quantity 50 μg
Source

Human embryonic kidney cell, HEK293-derived human NCAM-1/CD56 protein

Human NCAM-1/CD56 (Leu20-Gly708) Accession # P13591-1IEGRMDHuman IgG1 (Pro100-Lys330)N-terminus C-terminus

Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 2 weeks, 2 to 8°C under sterile conditions after reconstitution. 3 months, -20 to -70°C under sterile conditions after reconstitution.
Endotoxin Concentration <0.10 EU per 1 μg of the protein by the LAL method
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS
Buffer Lyophilized from a 0.2 μm filtered solution in PBS
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.