missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Recombinant Human Neuregulin-3/NRG3 Fc Chimera Protein, CF

Product Code. 18756165
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18756165 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18756165 Supplier R&D Systems™ Supplier No. 11373NR050
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Measured by its binding ability in a functional ELISA. Recombinant Human Neuregulin-3/NRG3 Fc Chimera (Catalog # 1373-NR) binds to Recombinant Human ErbB4/Her4 Fc Chimera Protein (Catalog # 1131-ER ) with a ED50 of 15.0-150 ng/mL.
TRUSTED_SUSTAINABILITY

Especificaciones

Accession Number P56975.1
For Use With (Application) Bioactivity
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
Gene ID (Entrez) 10718
Molecular Weight (g/mol) M.W. (Observed): 90-100 kDa, under reducing conditions., M.W. (Theroretical): 55 kDa
Quantity 50 μg
Source Human embryonic kidney cell, HEK293-derived human Neuregulin-3/NRG3 protein Human Neuregulin-3/NRG3 (Ser93-Arg359) (N-terminus) Accession # P56975.1 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Gene Alias HRG3, neuregulin 3, neuregulin 3 variant 1, neuregulin 3 variant 10, neuregulin 3 variant 11, neuregulin 3 variant 12, neuregulin 3 variant 13, neuregulin 3 variant 14, neuregulin 3 variant 3, neuregulin 3 variant 4, neuregulin 3 variant 5, neuregulin 3 variant 6, neuregulin 3 variant 7, neuregulin 3 variant 8, neuregulin 3 variant 9, neuregulin-3-like polypeptide, pro-neuregulin-3, membrane-bound isoform, pro-NRG3
Conjugate Unconjugated
Reconstitution Reconstitute at 500 μg/mL in PBS.
Product Line R&D Systems
Purity or Quality Grade >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Protein Neuregulin-3/NRG3
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.