Learn More
R&D Systems™ Recombinant Human Neurexin 2 alpha Isoform 2 Fc Chimera
Product Code. 18186820
Click to view available options
Quantity:
25 μg
Unit Size:
25µg
Product Code. 18186820
Brand: R&D Systems™ 6636NX025
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
The Recombinant Human Neurexin 2 alpha Isoform 2 Fc Chimera is derived from CHO. The Recombinant Human Neurexin 2 alpha Isoform 2 Fc Chimera has been validated for the following applications: Bioactivity.
TRUSTED_SUSTAINABILITY
Specifications
Formulation Lyophilized from a 0.2μm filtered solution in PBS.
Gene ID (Entrez) 9379
Molecular Weight (g/mol) 193 kDa (monomer)
Name Neurexin 2 alpha/NRXN2a Isoform 2 Fc Chimera
Quantity 25 μg
Source Chinese Hamster Ovary cell line,CHO-derived human Neurexin 2 alpha/NRXN2a protein, (N-terminus) Human Neurexin 2 alpha (Met1-Thr1566) Accession &Num; NP_620060 IEGRMD Human IgG1 (Pro100-Lys330) (C-terminus)
Storage Requirements Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution.
Structural Form Disulfide-linked homodimer
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade 95%, by SDS-PAGE under reducing conditions and visualized by silver stain.