missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Non-histone chromosomal protein HMG-17 Protein
A DNA sequence encoding the Homo sapiens (Human) Non-histone chromosomal protein HMG-17, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP6170-ec-1mg
Additional Details : Weight : 0.01000kg
Specifications
NP_005508.1 | |
Non-histone chromosomal protein HMG-17 Protein | |
Research Use Only | |
HMGN2 | |
Recombinant Protein | |
E. coli | |
PKRKAEGDAK GDKAKVKDEP QRRSARLSAK PAPPKPEPKP KKAPAKKGEK VPKGKKGKAD AGKEGNNPAE NGDAKTDQAQ KAEGAGDAK | |
Greater than 80% as determined by SDS-PAGE. |
3151 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |