missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human NT5C2 Protein
Click to view available options
Quantity:
1 mg
20 μg
5 μg
Unit Size:
1mg
20µg
5µg
Specifications
Specifications
| Name | NT5C2 Protein |
| Quantity | 20 μg |
| Regulatory Status | Research Use Only |
| Endotoxin Concentration | Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. |
| Product Type | Recombinant Protein |
| Cross Reactivity | Human |
| Species | E. coli |
| Protein Tag | His |
| Sequence | MGSSHHHHHH SSGLVPRGSH MSTSWSDRLQ NAADMPANMD KHALKKYRRE AYHRVFVNRS LAMEKIKCFG FDMDYTLAVY KSPEYESLGF ELTVERLVSI GYPQELLSFA YDSTFPTRGL VFDTLYGNLL KVDAYGNLLV CAHGFNFIRG PETREQYPNK FIQRDDTERF YILNTLFNLP ETYLLACLVD FFTNCPRYTS CETGFKDGDL FMSYRSMFQD VRDAVDWVHY KGSLKEKTVE NLEKYVVKDG KLPLLLSRMK EVGKVFLATN SDYKYTDKIM TYLFDFPHGP KPGSSHRPWQ SYFDLILVDA RKPLFFGEGT VLRQVDTKTG KLKIGTYTGP LQHGIVYSGG SSDTICDLLG AKGKDILYIG DHIFGDILKS KKRQGWRTFL VIPELAQELH VWTDKSSLFE ELQSLDIFLA ELYKHLDSSS NERPDISSIQ RRIKKVTHDM DMCYGMMGSL FRSGSRQTLF ASQVMRYADL YAASFINLLY YPFSYLFRAA HVLMPHESTV EHTHVDINEM ESPLATRNRT SVDFKDTDYK RHQLTRSISE IKPPNLFPLA PQEITHCHDE DDDEEEEEEE E |
| Buffer | NT5C2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 30% glycerol, 0.1M NaCl, 1mM DTT and 0.1mM PMSF. |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction